Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nNOS Antibody (60718), DyLight 680, Novus Biologicals™
SDP

Catalog No. MAB2416FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2416FR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2416FR Supplier Novus Biologicals Supplier No. MAB2416FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

nNOS Monoclonal specifically detects nNOS in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen nNOS
Applications Western Blot
Classification Monoclonal
Clone 60718
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bNOS, brain, Constitutive NOS, EC 1.14.13.39, IHPS1, Neuronal NOS, nitric oxide synthase 1 (neuronal), N-NOS, nNOSNOS type I, NOS, Peptidyl-cysteine S-nitrosylase NOS1
Gene Symbols NOS1
Host Species Mouse
Immunogen Partial recombinant human nNOS protein (amino acids 218-1434) [UniProt P29475]
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Asthma, Cancer, HIF Target Genes, Hypoxia, Immunology, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 4842
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.