Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

nNOS Antibody (85340), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB296798 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296798 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296798 Supplier Novus Biologicals Supplier No. FAB24161S

Mouse Monoclonal Antibody

nNOS Monoclonal antibody specifically detects nNOS in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry

Specifications

Antigen nNOS
Applications Western Blot, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 85340
Conjugate Alexa Fluor 750
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias bNOS, brain, Constitutive NOS, EC 1.14.13.39, IHPS1, Neuronal NOS, nitric oxide synthase 1 (neuronal), N-NOS, nNOSNOS type I, NOS, Peptidyl-cysteine S-nitrosylase NOS1
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human nNOS, Ser218-Ser1434, Accession # P29475
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Cancer, HIF Target Genes, Hypoxia, Immunology, Neuroscience, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4842
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.