Learn More
NNT1 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$250.00 - $2175.00
Specifications
Antigen | NNT1 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P18650UG
![]() |
Invitrogen™
500P18650UG |
50 μg |
Each for $250.00
|
|
|||||
500P1861MG
![]() |
Invitrogen™
500P1861MG |
1 mg |
Each for $2,175.00
|
|
|||||
Description
AA Sequence of recombinant protein: MLNRTGDPGP GPSIQKTYDL TRYLEHQLRS LAGTYLNYLG PPFNEPDFNP PRLGAETLPR ATVDLEVWRS LNDKLRLTQN YEAYSHLLCY LRGLNRQAAT AELRRSLAHF CTSLQGLLGS IAGVMAALGY PLPQPLPGTE PTWTPGPAHS DFLQKMDDFW LLKELQTWLW RSAKDFNRLK KKMQPPAAAV TLHLGAHGF. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human NNT-1/BCSF-3. Anti-Human NNT-1/BCSF-3-specific antibody was purified by affinity chromatography employing an immobilized Human NNT-1/BCSF-3 matrix. Sandwich ELISA: To detect Human NNT-1/BCSF-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human NNT-1/BCSF-3 (500-P186Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human NNT-1/BCSF-3. Western Blot: To detect Human NNT-1/BCSF-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human NNT-1/BCSF-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions
NNT-1/BCSF-3 is a neurotrophic factor with B-cell-stimulating capabilities. Expressed in the lymph nodes and spleen, NNT-1/BCSF-3 activates glycoprotein 130 (gp130) and leukemia inhibitory factor receptor member b (LIFR-b) by binding and inducing the tyrosine phosphorylation of these receptors. In vitro, it supports the survival of chick embryo motor and sympathetic neurons. In mice, NNT-1/BCSF-3 induces serum amyloid A, causes body weight loss, and B cell hyperplasia associated with increases in serum IgG and IgM.Specifications
NNT1 | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9UBD9 | |
23529 | |
E.coli-derived Recombinant Human NNT-1/BCSF-3. | |
Antigen Affinity Chromatography | |
CLCF1 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
b cell-stimulating factor 3; B-cell stimulating factor 3; B-cell stimulatory factor 3; B-cell-stimulating factor 3; Bsf3; BSF-3; Cardiotrophin-like cytokine factor 1; CISS2; Clc; CLCF1; CRLF1 associated cytokine-like factor 1; neurotrophin-1/B-cell stimulating factor-3; NNT1; NNT-1; NNT-1/BSF-3; Novel neurotrophin-1; NR6 | |
CLCF1 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.