Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOB1 Antibody (OTI1C12), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB273027A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB273027A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB273027A48 Supplier Novus Biologicals Supplier No. NBP273027AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NOB1 Monoclonal specifically detects NOB1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen NOB1
Applications Western Blot
Classification Monoclonal
Clone OTI1C12
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias adenocarcinoma antigen recognized by T lymphocytes 4, ART4, ART-4, MST158, nin one binding protein, NIN1/RPN12 binding protein 1 homolog (S. cerevisiae), NOB1PPSMD8BP1, Phosphorylation regulatory protein HP-10, Protein ART-4, PSMD8 binding protein 1, RNA-binding protein NOB1
Gene Symbols NOB1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 2-234 of human NOB1 (NP_054781) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 28987.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.