Learn More
Description
Specifications
Specifications
| Antigen | NOC3L |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml |
| Formulation | PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. | Q8WTT2 |
| Gene Alias | AD24NOC3-like protein, C10orf117, chromosome 10 open reading frame 117, Factor for adipocyte differentiation 24, FAD24NOC3 protein homolog, FLJ12820, nucleolar complex associated 3 homolog (S. cerevisiae), nucleolar complex protein 3 homolog, Nucleolar complex-associated protein 3-like protein |
| Gene Symbols | NOC3L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
