Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOC4L Antibody (PCRP-NOC4L-1E3), PerCP, Novus Biologicals™
SDP

Catalog No. NB381719 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381719 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381719 Supplier Novus Biologicals Supplier No. NBP308308PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NOC4L Monoclonal antibody specifically detects NOC4L in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen NOC4L
Applications Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-NOC4L-1E3
Conjugate PerCP
Formulation PBS
Gene Alias MGC3162, NET49, NOC4, NOC4 protein homolog, NOC4-like protein, nucleolar complex associated 4 homolog (S. cerevisiae), nucleolar complex protein 4 homolog, Nucleolar complex-associated protein 4-like protein
Host Species Mouse
Immunogen Recombinant full-length human NOC4L protein (Uniprot: Q9BVI4)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79050
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.