Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nocodazole, 98%, ACROS Organics™

Click to view available options
Quantity:
10mg
50mg
Packaging:
Glass bottle
31430-18-9
C14H11N3O3S
301.32
MFCD00005588
KYRVNWMVYQXFEU-UHFFFAOYSA-N
nocodazole,oncodazole,nocodazol,nocodazolum,nocidazole,nocodazole usan:inn,nocodazol inn-spanish,nocodazolum inn-latin,unii-sh1wy3r615,methyl 5-2-thenoyl-2-benzimidazolecarbamate
4122
CHEBI:34892
methyl N-[6-(thiophene-2-carbonyl)-1H-benzimidazol-2-yl]carbamate
COC(=O)NC1=NC2=C(N1)C=C(C=C2)C(=O)C3=CC=CS3
31430-18-9
C14H11N3O3S
301.32
MFCD00005588
KYRVNWMVYQXFEU-UHFFFAOYSA-N
nocodazole,oncodazole,nocodazol,nocodazolum,nocidazole,nocodazole usan:inn,nocodazol inn-spanish,nocodazolum inn-latin,unii-sh1wy3r615,methyl 5-2-thenoyl-2-benzimidazolecarbamate
4122
CHEBI:34892
methyl N-[6-(thiophene-2-carbonyl)-1H-benzimidazol-2-yl]carbamate
COC(=O)NC1=NC2=C(N1)C=C(C=C2)C(=O)C3=CC=CS3
Color Brown-Orange to White
Packaging Glass bottle
Quantity 10mg
Infrared Spectrum Authentic
Melting Point 300.0°C to 305.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmso,dmf,ethanol,chloroform,acetone
Formula Weight 301.32
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Nocodazole, 98%
Show More Show Less

GHS H Statement
Suspected of causing genetic defects.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Suspected of damaging the unborn child.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray

GHS Signal Word: Warning

EINECSNumber : 250-626-5

RTECSNumber : DD6521000