Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nocodazole, 98%

Catalog No. p-175686
Click to view available options
Quantity:
10 mg
50 mg
Packaging:
Glass bottle

CAS: 31430-18-9 | C14H11N3O3S | 301.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31430-18-9
Molecular Formula C14H11N3O3S
Molecular Weight (g/mol) 301.32
MDL Number MFCD00005588
InChI Key KYRVNWMVYQXFEU-UHFFFAOYSA-N
Synonym nocodazole, oncodazole, nocodazol, nocodazolum, nocidazole, nocodazole usan:inn, nocodazol inn-spanish, nocodazolum inn-latin, unii-sh1wy3r615, methyl 5-2-thenoyl-2-benzimidazolecarbamate
PubChem CID 4122
ChEBI CHEBI:34892
IUPAC Name methyl N-[6-(thiophene-2-carbonyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=C(N1)C=C(C=C2)C(=O)C3=CC=CS3

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 300.0°C to 305.0°C
Color Brown-Orange to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 mg
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmso,dmf,ethanol,chloroform,acetone
Formula Weight 301.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nocodazole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Suspected of causing genetic defects.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray
EINECSNumber 250-626-5
RTECSNumber DD6521000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.