Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOD1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$243.50 - $353.50

Specifications

Antigen NOD1
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056152T
SDP
View Documents
Novus Biologicals
NB10056152SS
0.025 mL
Each for $243.50
Only null left
Add to Cart
 
NB10056152
SDP
View Documents
Novus Biologicals
NB10056152
0.05 mL
Each for $353.50
Only null left
Add to Cart
 
Description

Description

NOD1 Polyclonal specifically detects NOD1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

NOD1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Apoptosis, Asthma, Autophagy, Immunology
Whole antisera with 0.05% Sodium Azide
10392
A synthetic peptide corresponding to amino acids 494-512 (QLGFLRALPELGPGGDQQS) of human Nod1/CARD4 was used as immunogen; GenBank no. gi 5174617 ref NP_006083.1 .
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot 1:1000-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Polyclonal
Antisera
RUO
Human
CARD4NLR family, CARD domain containing 1, caspase recruitment domain family, member 4, Caspase recruitment domain-containing protein 4, CLR7.1, NLRC1, nucleotide-binding oligomerization domain containing 1, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 1, nucleotide-binding oligomerization domain-containing protein 1
NOD1
IgG
Unpurified
NOD1
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.