Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOD2 Antibody (2D9) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 22 publications

Supplier:  Novus Biologicals NB100524

 View more versions of this product

Catalog No. NB100524


Only null left
Add to Cart

Description

Description

NOD2 Monoclonal specifically detects NOD2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications

Specifications

NOD2
Monoclonal
1.0 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry reported in scientific literature (PMID 28241127), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen, Flow (Intracellular)
BLAU, CARD15caspase recruitment domain protein 15, caspase recruitment domain family, member 15, Caspase recruitment domain-containing protein 15, CDACUG, CLR16.3, IBD1NLR family, CARD domain containing 2, Inflammatory bowel disease protein 1, NLRC2, NOD-like receptor C2, nucleotide-binding oligomerization domain 2, nucleotide-binding oligomerization domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 2, nucleotide-binding oligomerization domain-containing protein 2, PSORAS1NOD2B
Mouse
110 kDa
0.1 mL
Apoptosis, Autophagy, Cancer, Caspases, Cell Biology
64127
Human, Mouse
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
2D9
Unconjugated
Q9HC29
NOD2
Recombinant human NOD2 protein corresponding to residues 28-301. [UniProt# Q9HC29]
Protein G purified
RUO
Primary
Human and mouse. Immunogen has 92% homology to primate.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only