Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOD2 Antibody (2D9) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 22 publications

$269.00 - $562.50

Specifications

Antigen NOD2
Clone 2D9
Concentration 1.0 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry reported in scientific literature (PMID 28241127), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen, Flow (Intracellular)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB100524SS
SDP
View Documents
Novus Biologicals
NB100524SS
0.025 mL
Each for $269.00
Only null left
Add to Cart
 
NB100524
SDP
View Documents
Novus Biologicals
NB100524
0.1 mL
Each for $562.50
Only null left
Add to Cart
 
Description

Description

NOD2 Monoclonal specifically detects NOD2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications

Specifications

NOD2
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Apoptosis, Autophagy, Cancer, Caspases, Cell Biology
Q9HC29
64127
Recombinant human NOD2 protein corresponding to residues 28-301. [UniProt# Q9HC29]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
110 kDa
2D9
Western Blot 1:100-1:2000, Flow Cytometry reported in scientific literature (PMID 28241127), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen, Flow (Intracellular)
Monoclonal
Purified
RUO
Human, Mouse
BLAU, CARD15caspase recruitment domain protein 15, caspase recruitment domain family, member 15, Caspase recruitment domain-containing protein 15, CDACUG, CLR16.3, IBD1NLR family, CARD domain containing 2, Inflammatory bowel disease protein 1, NLRC2, NOD-like receptor C2, nucleotide-binding oligomerization domain 2, nucleotide-binding oligomerization domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 2, nucleotide-binding oligomerization domain-containing protein 2, PSORAS1NOD2B
NOD2
IgG1
Protein G purified
Human and mouse. Immunogen has 92% homology to primate.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.