Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOD2 Antibody (2D9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 22 publications
Supplier: Novus Biologicals NB100524SS
Description
NOD2 Monoclonal specifically detects NOD2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).Specifications
NOD2 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry reported in scientific literature (PMID 28241127), Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen, Flow (Intracellular) | |
BLAU, CARD15caspase recruitment domain protein 15, caspase recruitment domain family, member 15, Caspase recruitment domain-containing protein 15, CDACUG, CLR16.3, IBD1NLR family, CARD domain containing 2, Inflammatory bowel disease protein 1, NLRC2, NOD-like receptor C2, nucleotide-binding oligomerization domain 2, nucleotide-binding oligomerization domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 2, nucleotide-binding oligomerization domain-containing protein 2, PSORAS1NOD2B | |
Mouse | |
110 kDa | |
0.025 mL | |
Apoptosis, Autophagy, Cancer, Caspases, Cell Biology | |
64127 | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
2D9 | |
Unconjugated | |
Q9HC29 | |
NOD2 | |
Recombinant human NOD2 protein corresponding to residues 28-301. [UniProt# Q9HC29] | |
Protein G purified | |
RUO | |
Primary | |
Human and mouse. Immunogen has 92% homology to primate. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction