Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOD2 Antibody (2D9), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB100524R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100524R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100524R Supplier Novus Biologicals Supplier No. NB100524R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NOD2 Monoclonal specifically detects NOD2 in Human, Mouse samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen NOD2
Applications Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2D9
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
Gene Accession No. Q9HC29
Gene Alias BLAU, CARD15caspase recruitment domain protein 15, caspase recruitment domain family, member 15, Caspase recruitment domain-containing protein 15, CDACUG, CLR16.3, IBD1NLR family, CARD domain containing 2, Inflammatory bowel disease protein 1, NLRC2, NOD-like receptor C2, nucleotide-binding oligomerization domain 2, nucleotide-binding oligomerization domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 2, nucleotide-binding oligomerization domain-containing protein 2, PSORAS1NOD2B
Gene Symbols NOD2
Host Species Mouse
Immunogen Recombinant human NOD2 protein corresponding to residues 28-301. [UniProt# Q9HC29]
Molecular Weight of Antigen 110 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Caspases, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 64127
Test Specificity Human and mouse. Immunogen has 92% homology to primate.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.