Learn More
Description
Specifications
Specifications
| Antigen | NOD2 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000∼, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000∼, Immunohistochemistry-Frozen 1:1000-1:5000 |
| Formulation | 50 ul neat whole antisera with 0.05% Sodium Azide |
| Gene Accession No. | Q9HC29 |
| Gene Alias | BLAU, CARD15caspase recruitment domain protein 15, caspase recruitment domain family, member 15, Caspase recruitment domain-containing protein 15, CDACUG, CLR16.3, IBD1NLR family, CARD domain containing 2, Inflammatory bowel disease protein 1, NLRC2, NOD-like receptor C2, nucleotide-binding oligomerization domain 2, nucleotide-binding oligomerization domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 2, nucleotide-binding oligomerization domain-containing protein 2, PSORAS1NOD2B |
| Gene Symbols | NOD2 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
