Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NOD2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$243.50 - $353.50
Specifications
Antigen | NOD2 |
---|---|
Dilution | Western Blot 1:1000-1:2000∼, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000∼, Immunohistochemistry-Frozen 1:1000-1:5000 |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP227328SS
![]() |
Novus Biologicals
NBP227328SS |
0.025 mL |
Each for $243.50
|
|
|||||
NBP227328
![]() |
Novus Biologicals
NBP227328 |
0.05 mL |
Each for $353.50
|
|
|||||
Description
NOD2 Polyclonal specifically detects NOD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
NOD2 | |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
Rabbit | |
Apoptosis, Autophagy, Cancer, Caspases, Cell Biology | |
50 ul neat whole antisera with 0.05% Sodium Azide | |
BLAU, CARD15caspase recruitment domain protein 15, caspase recruitment domain family, member 15, Caspase recruitment domain-containing protein 15, CDACUG, CLR16.3, IBD1NLR family, CARD domain containing 2, Inflammatory bowel disease protein 1, NLRC2, NOD-like receptor C2, nucleotide-binding oligomerization domain 2, nucleotide-binding oligomerization domain containing 2, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 2, nucleotide-binding oligomerization domain-containing protein 2, PSORAS1NOD2B | |
NOD2 | |
IgG | |
Unpurified |
Western Blot 1:1000-1:2000∼, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000∼, Immunohistochemistry-Frozen 1:1000-1:5000 | |
Polyclonal | |
Antisera | |
RUO | |
Human | |
Q9HC29 | |
64127 | |
A synthetic peptide corresponding to amino acids 582-610 of human NOD2/CARD15 was used as immunogen; GenBank no. NP_071445 NOD2 is a 1040 amino acid protein. The immunogen sequence is 100% conserved in certain NOD2 isoforms and mutations. The immunogen sequence corresponds to amino acids 45-63 of a short form of NOD2. This NOD2 short form is 273 amino acids; GenBank no.AAk70868.1 aa 582-600; N-Terminus; human | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title