Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Noggin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10093547 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093547 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093547 Supplier Novus Biologicals Supplier No. NB10093547
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Noggin Polyclonal specifically detects Noggin in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Noggin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q13253
Gene Alias noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1
Gene Symbols NOG
Host Species Rabbit
Immunogen Synthetic peptide comprising an internal sequence of the human Noggin protein. Sequence is 100% conserved in rat and mouse (NP_005441).
Molecular Weight of Antigen 25.7 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9241
Test Specificity Noggin
Target Species Human, Mouse, Rat, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.