Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nogo Antibody, Novus Biologicals™
SDP

Catalog No. NB10056681 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056681 0.2 mL
NB10056681T 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056681 Supplier Novus Biologicals Supplier No. NB10056681
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 16 publications

Nogo Polyclonal specifically detects Nogo in Human, Mouse, Ovine, Rabbit, Rat, Sheep samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot.

Specifications

Antigen Nogo
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Simple Western 1:1000, ELISA reported in scientific literature (PMID 17413036), Immunohistochemistry reported in scientific literature (PMID 30208932), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27132996), Immunohistochemistry-Paraffin 1:100
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. Q9NQC3
Gene Alias ASY, foocen, Human NogoA, KIAA0886, My043 protein, Nbla00271, Nbla10545, neurite growth inhibitor 220, Neurite outgrowth inhibitor, Neuroendocrine-specific protein, Neuroendocrine-specific protein C homolog, NI220/250, Nogo protein, NOGO-A, Nogo-B, Nogo-C, NOGOC, NOGORTN4-B1, NSP, NSP-CL, reticulon 4, reticulon 5, reticulon-4, reticulon-5, RTN4-A, RTN4-B2, RTN4-C, RTN-x
Gene Symbols RTN4
Host Species Rabbit
Immunogen A synthetic peptide to an internal portion of human Nogo A (between amino acids 1-50) [UniProt Q9NQC3].
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 57142
Test Specificity This recognizes Nogo-A (approx. 220 kD)and Nogo-B2 and B1 (approx. 50 kD and 48 kD in human and 43 kD and 41 kD in mouse and rat, respectively).
Target Species Human, Mouse, Mouse, Ovine, Rabbit, Rat, Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.