Learn More
Description
Specifications
Specifications
| Antigen | Nogo |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Simple Western, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Simple Western 1:1000, ELISA reported in scientific literature (PMID 17413036), Immunohistochemistry reported in scientific literature (PMID 30208932), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27132996), Immunohistochemistry-Paraffin 1:100 |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | Q9NQC3 |
| Gene Alias | ASY, foocen, Human NogoA, KIAA0886, My043 protein, Nbla00271, Nbla10545, neurite growth inhibitor 220, Neurite outgrowth inhibitor, Neuroendocrine-specific protein, Neuroendocrine-specific protein C homolog, NI220/250, Nogo protein, NOGO-A, Nogo-B, Nogo-C, NOGOC, NOGORTN4-B1, NSP, NSP-CL, reticulon 4, reticulon 5, reticulon-4, reticulon-5, RTN4-A, RTN4-B2, RTN4-C, RTN-x |
| Gene Symbols | RTN4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
