Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nogo B receptor Antibody, Novus Biologicals™
SDP

Catalog No. NBP224950 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224950 0.1 mg
NBP224950SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224950 Supplier Novus Biologicals Supplier No. NBP224950
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Nogo B receptor Polyclonal specifically detects Nogo B receptor in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Nogo B receptor
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias C6orf68MGC117249, chromosome 6 open reading frame 68, MGC:7199, MGC7199, NgBR, nogo-B receptor, Nuclear undecaprenyl pyrophosphate synthase 1 homolog, nuclear undecaprenyl pyrophosphate synthase 1 homolog (S. cerevisiae)
Gene Symbols NUS1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 64-74 (RNRRHHRHPRG) of human Nogo-B receptor (NgBR) was used as immunogen, GenBank no sp Q96E22.1 NGBR_HUMAN (Miao et al, 2006). This immunogen sequence is within the 93 amino acid putative ectodomain of the NgBR.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 116150
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.