Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nogo Receptor/NgR Antibody (M5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP25267525 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25267525 0.025 mg
NBP25267502 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP25267525 Supplier Novus Biologicals Supplier No. NBP2526750.025MG
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Nogo Receptor/NgR Monoclonal specifically detects Nogo Receptor/NgR in Human, Mouse, Rat samples. It is validated for CyTOF, ELISA, Flow Cytometry, Immunoprecipitation, Western Blot.

Specifications

Antigen Nogo Receptor/NgR
Applications CyTOF, ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M5
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready
Formulation PBS with 0.02% Proclin 300. with 0.02% Proclin 300
Gene Alias NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor
Gene Symbols RTN4R
Host Species Rabbit
Immunogen (His)6-tagged fragment of rat NgR1 (amino acids 278-473).
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65078
Test Specificity The antibody binds to human and rat NgR1 with a Kd of 2.9 nM for both species (bivalent binding).
Target Species Human, Mouse, Rat
Content And Storage Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.