Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nogo Receptor/NgR Antibody (M5) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $608.50
Specifications
| Antigen | Nogo Receptor/NgR |
|---|---|
| Clone | M5 |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25267525
![]() |
Novus Biologicals
NBP2526750.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NBP25267502
![]() |
Novus Biologicals
NBP2526750.2MG |
0.2 mg |
Each for $608.50
|
|
|||||
Description
Nogo Receptor/NgR Monoclonal specifically detects Nogo Receptor/NgR in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF-ready.Specifications
| Nogo Receptor/NgR | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| 65078 | |
| (His)6-tagged fragment of rat NgR1 (amino acids 278-473). | |
| Primary | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
| M5 | |
| Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor | |
| RTN4R | |
| IgG | |
| Protein A or G purified | |
| The antibody binds to human and rat NgR1 with a Kd of 2.9 nM for both species (bivalent binding). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title