Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nogo Receptor/NgR Antibody (M5), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB363045 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363045 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363045 Supplier Novus Biologicals Supplier No. NBP252675MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Nogo Receptor/NgR Monoclonal antibody specifically detects Nogo Receptor/NgR in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF

Specifications

Antigen Nogo Receptor/NgR
Applications Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF
Classification Monoclonal
Clone M5
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, ELISA, Immunoprecipitation, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor
Host Species Rabbit
Immunogen (His)6-tagged fragment of rat NgR1 (amino acids 278-473).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 65078
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.