Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nogo Receptor/NgR Antibody (MM0493-6H43) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB018912 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018912 0.025 mg
NBP211823 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018912 Supplier Novus Biologicals Supplier No. NBP2118230.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nogo Receptor/NgR Monoclonal specifically detects Nogo Receptor/NgR in Human samples. It is validated for Western Blot.

Specifications

Antigen Nogo Receptor/NgR
Applications Western Blot
Classification Monoclonal
Clone MM0493-6H43
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation A 0.2 um filtered solution in PBS with No Preservative
Gene Accession No. Q9BZR6
Gene Alias NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor
Gene Symbols RTN4R
Host Species Mouse
Immunogen Human Nogo Receptor
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65078
Reconstitution Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.