Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nogo Receptor/NgR Antibody (MM0493-6H43), HRP, Novus Biologicals™
SDP

Catalog No. NBP211823H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211823H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211823H Supplier Novus Biologicals Supplier No. NBP211823H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Nogo Receptor/NgR Monoclonal specifically detects Nogo Receptor/NgR in Human samples. It is validated for Western Blot.

Specifications

Antigen Nogo Receptor/NgR
Applications Western Blot
Classification Monoclonal
Clone MM0493-6H43
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias NgR, Nogo receptor, Nogo-66 receptor, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor
Gene Symbols RTN4R
Host Species Mouse
Immunogen Human recombinant Nogo Receptor/NgR (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 65078
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.