Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOL1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP192192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP192192 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP192192 Supplier Novus Biologicals Supplier No. NBP192192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

NOL1 Polyclonal specifically detects NOL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen NOL1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P46087
Gene Alias EC 2.1.1, EC 2.1.1.-, MGC149287, MGC149288, NOL1/NOP2/Sun domain family, member 1, NOL1MGC117384, NOP120, NOP2 nucleolar protein homolog (yeast), NOP2/Sun domain family, member 1, NSUN1, Nucleolar protein 1, nucleolar protein 1 (120kD), nucleolar protein 1, 120kDa, Nucleolar protein 2 homolog, nucleolar protein 2 homolog (yeast), p120, Proliferating-cell nucleolar antigen p120, Proliferation-associated nucleolar protein p120, putative ribosomal RNA methyltransferase NOP2
Gene Symbols NOP2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:KLMDLFPLSELVEFLEANEVPRPVTLRTNTLKTRRRDLAQALINRGVNLDPLGKWSKTGLVVYDSSVPIGATPEYLAGH
Molecular Weight of Antigen 89 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4839
Test Specificity Specificity of human NOL1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.