Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOLC1 Antibody, Novus Biologicals™
SDP

Catalog No. NB404650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB404650 25 μL
NBP238625 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB404650 Supplier Novus Biologicals Supplier No. NBP23862525UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NOLC1 Polyclonal specifically detects NOLC1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NOLC1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q14978
Gene Alias 140 kDa nucleolar phosphoprotein, HCV NS5A trans-regulated protein 13, HCV NS5A-transactivated protein 13, Hepatitis C virus NS5A-transactivated protein 13, KIAA0035NS5ATP13, NOPP130, NOPP140, Nucleolar 130 kDa protein, nucleolar and coiled-body phosphoprotein 1, nucleolar and coiled-body phosphprotein 1, Nucleolar phosphoprotein p130, nucleolar protein p130, P130
Gene Symbols NOLC1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: VVVSKSGSLKKRKQNEAAKEAETPQAKKIKLQTPNTFPKRKKGEKRASSPFRRVREEEIEVDSRVADNSFDAKRGAAGDWGERANQVLKF
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9221
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.