Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Nomex™ / Lenzing FR Full Face Hood

Catalog No. p-7180093 Shop All Protective Industrial Products Inc. Products
Click to view available options
Color:
Black
White
Design Type:
Straight Cut Design
Tri-Cut Design
This item is not returnable. View return policy
This item is not returnable. View return policy

PIP™ Nomex™ / Lenzing FR Full Face Hood features a fabric combination that provides comfort and breathability

Specifications

Product Type Fire Protectant Hood
Size Universal
Color White
Disposable Reusable
Design Type Tri-Cut Design
Certifications/Compliance Meets NFPA 1971 Requirements, TPP Rating 23
Quantity 1 Ea.
Includes Hood Only
For Use With (Application) Heat/Fire Applications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.