Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

non-muscle heavy chain 10 Myosin Antibody, Novus Biologicals™
SDP

Catalog No. NBP118931 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP118931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP118931 Supplier Novus Biologicals Supplier No. NBP118931
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

non-muscle heavy chain 10 Myosin Polyclonal specifically detects non-muscle heavy chain 10 Myosin in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen non-muscle heavy chain 10 Myosin
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 1:10-1:500
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_005955.1
Gene Alias Cellular myosin heavy chain, type B, cellular myosin heavy chain, type B type B, heavy polypeptide 10, non-muscle, MGC134913, MGC134914, Myosin heavy chain 10, Myosin heavy chain, non-muscle IIb, myosin heavy chain, nonmuscle type B, myosin, heavy chain 10, non-muscle, myosin-10, near to the ATP binding region, NMMHC II-b, NMMHCB, NMMHC-B, NMMHC-IIB, Non-muscle myosin heavy chain B, nonmuscle myosin heavy chain IIB, Non-muscle myosin heavy chain IIb, nonmuscle myosin heavy chain-B, nonmuscle myosin II heavy chain-B
Gene Symbols MYH10
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1935 and 1955 of human myosin, heavy chain 10, non-muscle using the numbering given in entry NP_005955.1 (GeneID 4628).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4628
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.