Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

non-muscle heavy chain 10 Myosin Antibody, Novus Biologicals™
SDP

Catalog No. NB436176 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB436176 25 μL
NBP238566 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB436176 Supplier Novus Biologicals Supplier No. NBP23856625UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

non-muscle heavy chain 10 Myosin Polyclonal specifically detects non-muscle heavy chain 10 Myosin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen non-muscle heavy chain 10 Myosin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. P35580
Gene Alias Cellular myosin heavy chain, type B, cellular myosin heavy chain, type B type B, heavy polypeptide 10, non-muscle, MGC134913, MGC134914, Myosin heavy chain 10, Myosin heavy chain, non-muscle IIb, myosin heavy chain, nonmuscle type B, myosin, heavy chain 10, non-muscle, myosin-10, near to the ATP binding region, NMMHC II-b, NMMHCB, NMMHC-B, NMMHC-IIB, Non-muscle myosin heavy chain B, nonmuscle myosin heavy chain IIB, Non-muscle myosin heavy chain IIb, nonmuscle myosin heavy chain-B, nonmuscle myosin II heavy chain-B
Gene Symbols MYH10
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: RQLDDATEANEGLSREVSTLKNRLRRGGPISFSSSRSGRRQLHLEGASLELSDDDTESKTSDVNETQPPQ
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4628
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.