Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

non-muscle Myosin IIA, p Ser1943 Antibody, Novus Biologicals™
SDP

Catalog No. NB387829 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387829 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387829 Supplier Novus Biologicals Supplier No. NBP323432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

non-muscle Myosin IIA Polyclonal antibody specifically detects non-muscle Myosin IIA in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen non-muscle Myosin IIA
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/ Immunofluorescence 1:100
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias Cellular myosin heavy chain, type A, DFNA17, EPSTS, FTNSNMMHC-IIA, MGC104539, MHANMMHC II-a, MYH9 variant protein, Myosin heavy chain 9, Myosin heavy chain, non-muscle IIa, myosin, heavy chain 9, non-muscle, myosin, heavy polypeptide 9, non-muscle, myosin-9, NMHC-II-A, NMMHCA, NMMHC-A, Non-muscle myosin heavy chain A, nonmuscle myosin heavy chain II-A, Non-muscle myosin heavy chain IIa, non-muscle myosin heavy polypeptide 9
Host Species Rabbit
Immunogen Phospho-non-muscle Myosin IIA (Ser-1943) synthetic peptide (coupled to KLH) corresponding to amino acid residues surrounding Ser-1943 in human myosin IIA heavy chain. This peptide sequence is highly conserved in rat and mouse non-muscle Myosin IIA, but is not found in non-muscle Myosin IIB and is not well conserved in non-muscle Myosin IIC. [UniProt# P35579]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 4627
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.