Learn More
Description
Specifications
Specifications
| Assay Range | 1.875 - 60 ng/ml |
| Assay Sensitivity | 0.59 ng/ml |
| Cross Reactivity | The DNA Damage ELISA also detects 8-hydroxyguanosine (product of oxidative RNA damage) and 8-hydroxyguanine (product of oxidative DNA damage by hydroxyl radicals). |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Cell Lysates, Cell Culture Supernatant, Plasma, Saliva, Seminal Fluids, Serum, Urine |
| For Use With (Application) | ELISA |
| Host Species | Non-species specific |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Target | 8-OHdG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
