Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonadecane, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
629-92-5
C19H40
268.51
MFCD00009012
LQERIDTXQFOHKA-UHFFFAOYSA-N
n-nonadecane,nonadekan,unii-nmy21d3y5t,nmy21d3y5t,unknown branched fragment of phospholipid,ch3-ch2 17-ch3,unknown phospholipid fragment,nonadecane, analytical standard,upl
12401
CHEBI:32927
nonadecane
CCCCCCCCCCCCCCCCCCC
629-92-5
C19H40
268.51
MFCD00009012
LQERIDTXQFOHKA-UHFFFAOYSA-N
n-nonadecane,nonadekan,unii-nmy21d3y5t,nmy21d3y5t,unknown branched fragment of phospholipid,ch3-ch2 17-ch3,unknown phospholipid fragment,nonadecane, analytical standard,upl
12401
CHEBI:32927
nonadecane
CCCCCCCCCCCCCCCCCCC
Viscosity 3.6 mPa.s (40°C)
Infrared Spectrum Authentic
Boiling Point 330.0°C
Color White
Packaging Glass bottle
Flash Point 168°C
Melting Point 32.0°C to 34.0°C
Quantity 25g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3(CH2)17CH3
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 268.51
Physical Form Crystalline Solid
Percent Purity 99%
Chemical Name or Material Nonadecane
Show More Show Less

GHS P Statement May be fatal if swallowed and enters airways. Repeated exposure may cause skin dryness or cracking.

GHS P Statement IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting.

Danger