Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NONO Antibody (OTI3F8), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP273033R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273033R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273033R Supplier Novus Biologicals Supplier No. NBP273033R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NONO Monoclonal specifically detects NONO in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NONO
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI3F8
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias NMT5552 kDa subunit, non-POU domain containing, octamer-binding, NRB54non-POU-domain-containing, octamer-binding, p54(nrb)
Gene Symbols NONO
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 184-385 of human NONO (NP_031389) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4841.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.