Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOP10 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen NOP10
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP156381
SDP
View Documents
Novus Biologicals
NBP156381
100 μL
Each for $487.50
Only null left
Add to Cart
 
NBP1563820
SDP
View Documents
Novus Biologicals
NBP15638120UL
20 μL N/A N/A N/A
Description

Description

NOP10 Polyclonal specifically detects NOP10 in Human samples. It is validated for Western Blot.
Specifications

Specifications

NOP10
Polyclonal
Rabbit
Core ESC Like Genes, Stem Cell Markers
H/ACA ribonucleoprotein complex subunit 3, homolog of yeast Nop10p, MGC70651, NOLA3Nucleolar protein family A member 3, NOP10 ribonucleoprotein homolog (yeast), NOP10P, Nucleolar protein 10, nucleolar protein family A, member 3 (H/ACA small nucleolar RNPs), snoRNP protein NOP10
NOP10
IgG
This product is specific to Subunit or Isoform: 3.
Western Blot
Unconjugated
RUO
Q9NPE3
55505
Synthetic peptides corresponding to NOLA3 The peptide sequence was selected from the middle region of NOLA3. Peptide sequence MFLQYYLNEQGDRVYTLKKFDPMGQQTCSAHPARFSPDDKYSRHRITIKK.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.