Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOP10 Antibody (SR1053), Novus Biologicals™
SDP

Catalog No. NB258435 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258435 100 μg
NB259432 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258435 Supplier Novus Biologicals Supplier No. NBP322490100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

NOP10 Monoclonal antibody specifically detects NOP10 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen NOP10
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone SR1053
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias H/ACA ribonucleoprotein complex subunit 3, homolog of yeast Nop10p, MGC70651, NOLA3Nucleolar protein family A member 3, NOP10 ribonucleoprotein homolog (yeast), NOP10P, Nucleolar protein 10, nucleolar protein family A, member 3 (H/ACA small nucleolar RNPs), snoRNP protein NOP10
Host Species Rabbit
Immunogen A synthesized peptide derived from human NOP10 (Uniprot #: Q9NPE3)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 55505
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.