Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOP58 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146846 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146846 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146846 Supplier Novus Biologicals Supplier No. NBP146846
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NOP58 Polyclonal specifically detects NOP58 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen NOP58
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:200- 1:1000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:1000
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Alias HSPC120, NOL5, NOP5/NOP58, NOP58 ribonucleoprotein homolog (yeast), NOP5nucleolar protein 58, Nucleolar protein 5, nucleolar protein NOP5/NOP58
Gene Symbols NOP58
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 479 and 529 of human NOP58 ribonucleoprotein homolog using the numbering given in entry NP_057018.1 (GeneID 51602).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research
Primary or Secondary Primary
Gene ID (Entrez) 51602
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Red flour beetle.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.