Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nope/IGDCC4 Antibody (2516A), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB340077 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340077 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340077 Supplier Novus Biologicals Supplier No. FAB10345MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Nope/IGDCC4 Monoclonal specifically detects Nope/IGDCC4 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot.

Specifications

Antigen Nope/IGDCC4
Applications CyTOF, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2516A
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias DDM36, hDDM36, immunoglobulin superfamily DCC subclass member 4, immunoglobulin superfamily, DCC subclass, member 4, KIAA1628, likely ortholog of mouse neighbor of Punc E11, LOC57722, Neighbor of punc e11, NOPEFLJ42051, Protein DDM36
Host Species Rabbit
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Nope/IGDCC4, Met1-Thr957, Accession # Q8TDY8
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57722
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.