Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NORE1 Antibody (OTI1C6), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP273039A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273039A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273039A32 Supplier Novus Biologicals Supplier No. NBP273039AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NORE1 Monoclonal specifically detects NORE1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen NORE1
Applications Western Blot
Classification Monoclonal
Clone OTI1C6
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Maxp1, MGC10823, MGC17344, New ras effector 1, NORE1A, NORE1B, NORE1RASSF3, Rap1-binding protein, RAPLnovel Ras effector 1, Ras association (RalGDS/AF-6) domain family 5, Ras association (RalGDS/AF-6) domain family member 5, ras association domain-containing protein 5, Ras effector-like protein, regulator for cell adhesion and polarization enriched in lymphoid tissue, Regulator for cell adhesion and polarization enriched in lymphoid tissues, tumor suppressor RASSF3
Gene Symbols RASSF5
Host Species Mouse
Immunogen Full length recombinant protein of human NORE1 (NP_872606) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 83593.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.