Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Norfloxacin

Catalog No. p-7042473
Click to view available options
Quantity:
1 g
5 g
25 g

A synthetic fluoroquinolone antibiotic | CAS: 70458-96-7 | C16H18FN3O3 | 319.336 g/mol

It finds it application as a fluorinated quinolone antibacterial. It is clinically used to treat urinary tract infections and prostatitis. In neutrophils from cirrhotic subjects, norfloxacin increases expression of IL-10 and heme oxygenase 1 (HO-1) and decreases expression of pro-inflammatory cytokines. Additionally, when complexed with gold(III), norfloxacin binds DNA and inhibits cellular proliferation in several cancer cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in acetic acid. Also soluble in acetone or cloroform. Slightly soluble in water

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70458-96-7
Molecular Formula C16H18FN3O3
Molecular Weight (g/mol) 319.336
MDL Number MFCD00079532
InChI Key OGJPXUAPXNRGGI-UHFFFAOYSA-N
Synonym Baccidal; Sebercim
PubChem CID 4539
ChEBI CHEBI:100246
IUPAC Name 1-ethyl-6-fluoro-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid
SMILES CCN1C=C(C(=O)C2=CC(=C(C=C21)N3CCNCC3)F)C(=O)O

Specifications

Type Norfloxacin
Physical Form Powder
Melting Point 220°C to 221°C
Color White to Yellow
Quantity 1 g
Merck Index 14,6700
Solubility Information Soluble in acetic acid; Also soluble in acetone or cloroform; Slightly soluble in water
Formula Weight 319.33
Chemical Name or Material Norfloxacin
EINECSNumber 274-614-4
RTECSNumber VB5005000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.