Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Norovirus (MNV-1) Mouse anti-Virus, Unconjugated, Clone: 5C4.10, MilliporeSigma™
Mouse Monoclonal Antibody
$207.00 - $541.00
Specifications
Antigen | Norovirus (MNV-1) |
---|---|
Clone | 5C4.10 |
Concentration | Please refer to lot specific datasheet. |
Applications | ELISA, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF2097-25
|
MilliporeSigma
MABF209725UG |
25 μg |
Each for $207.00
|
|
|||||
MABF2097-100
|
MilliporeSigma
MABF2097100UG |
100 μg |
Each for $541.00
|
|
|||||
Description
Anti-Norovirus (MNV-1), clone 5C4.10, Cat. No. MABF2097, is a mouse monoclonal antibody that detects murine Norovirus capsid protein and has been tested for use in ELISA and Western Blotting.Specifications
Norovirus (MNV-1) | |
Please refer to lot specific datasheet. | |
Monoclonal | |
Purified | |
RUO | |
Virus | |
MNV-1;Murine Norovirus | |
IgG2b κ | |
Protein G purified | |
Clone 5C4.10 specifically recognizes the S domain portion of capsid protein of murine Norovisrus. | |
∽57 kDa observed. Uncharacterized bands may be observed in some lysate(s). |
5C4.10 | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Inflammation & Immunology | |
Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
Infectious and UV-inactivated murine norovirus-1. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title