Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Norovirus VP1 Antibody (HL2410) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB295037 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295037 Supplier Novus Biologicals Supplier No. NBP325580
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Norovirus VP1 Monoclonal antibody specifically detects Norovirus VP1 in Virus samples. It is validated for Western Blot

Specifications

Antigen Norovirus VP1
Applications Western Blot
Classification Monoclonal
Clone HL2410
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000
Formulation PBS
Gene Alias Capsid Protein, Capsid protein VP1, CP, Norovirus Capsid protein VP1, Norwalk virus VP1, NoV VP1, p59
Host Species Rabbit
Immunogen Synthetic peptide of Norovirus VP1.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.