Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ 5500 Series Convenience Pack Respirators

Elastomeric half mask respirators offer a wide contoured sealing with a secure fit. Choose from several filter options depending on specific application.

Specifications

Certifications (NIOSH) NIOSH 42 CFR Part 84
Color Black
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Facepiece Material Thermoplastic Elastomer
View More Specs

Products 18
Catalog Number Mfr. No. Cartridge Type Size Includes Price Quantity & Availability  
Catalog Number Mfr. No. Cartridge Type Size Includes Price Quantity & Availability  
19-999-216
View Documents
Honeywell™
5501S
Organic Vapor Small OV Cartridge N/A N/A N/A
19-999-209
View Documents
Honeywell™
5501M
Organic Vapor Medium OV Cartridge N/A N/A N/A
19-999-217
View Documents
Honeywell™
5503L
Organic Vapor, Acid Gas Large OV and AG Cartridges N/A N/A N/A
19-999-213
View Documents
Honeywell™
5501N95M
Organic Vapor Medium OV Cartridge, N95 Filter N/A N/A N/A
19-999-210
View Documents
Honeywell™
5501N95L
Organic Vapor Large OV Cartridge, N95 Filter N/A N/A N/A
19-999-214
View Documents
Honeywell™
5501R95M
Organic Vapor Medium OV Cartridge, R95 Filter N/A N/A N/A
19-999-223
View Documents
Honeywell™
5580P100S
P100 Cartridge Small P100 Particulate Filters N/A N/A N/A
19-999-222
View Documents
Honeywell™
5580P100M
P100 Cartridge Medium P100 Particulate Filters N/A N/A N/A
19-999-221
View Documents
Honeywell™
5580P100L
P100 Cartridge Large P100 Particulate Filters N/A N/A N/A
19-999-226
View Documents
Honeywell™
5581P100S
N/A N/A N/A
19-999-225
View Documents
Honeywell™
5581P100M
Organic Vapor Medium OV Cartridge, P100 Particulate Filters N/A N/A N/A
19-999-224
View Documents
Honeywell™
5581P100L
N/A N/A N/A
19-999-229
View Documents
Honeywell™
5583P100S
N/A N/A N/A
19-999-228
View Documents
Honeywell™
5583P100M
Organic Vapor, Acid Gas Medium OV and AG Cartridges, P100 Particulate Filters N/A N/A N/A
19-999-227
View Documents
Honeywell™
5583P100L
N/A N/A N/A
19-999-207
View Documents
Honeywell™
55SCP100S
Defender™ Multi-Contaminant Cartridges Small Multi-Contaminant Cartridges and P100 Filters N/A N/A N/A
19-999-205
View Documents
Honeywell™
55SCP100M
Defender™ Multi-Contaminant Cartridges Medium Multi-Contaminant Cartridges and P100 Filters N/A N/A N/A
19-999-206
View Documents
Honeywell™
55SCP100L
Defender™ Multi-Contaminant Cartridges Large Multi-Contaminant Cartridges and P100 Filters N/A N/A N/A
Description

Description

  • Low-maintenance, reusable, economical
  • Soft, comfortable elastomer facepiece
  • For use with North cartridges, filters and accessories
  • Incorporates the anatomic design of the North 7700 Series respirators
  • Requires very little maintenance
  • Contoured sealing flange and cradle suspension system combine to ensure proper positioning
  • All parts are replaceable, extending service life
  • Easy-to-order 5500 Series Convenience Packs include most commonly used cartridges and filters
  • Available in a range of sizes and cartridge/filter combinations
Specifications

Specifications

NIOSH 42 CFR Part 84
Thermoplastic Elastomer
Thermoplastic Elastomer
Black
Half-Mask Respirator
Agriculture, food services, government, municipal service
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.