Learn More
Honeywell™ North™ BP1000 Series Backpack Air-Purifying Respirator
Convert your half mask or full facepiece to a welding respirator with this welding attachment. Puts cartridges on the back, increasing service life and minimizing weight to the facepiece.
Specifications
Certifications (NIOSH) | NIOSH 42 CFR Part 84 |
---|---|
For Use With (Application) | Welding and other high-heat operations |
For Use With (Equipment) | North™ 5500, 7700, 5400 and 7600 Series facepieces |
Includes | Backpack adapter |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-040-889A
|
Honeywell™
BP1201S |
Small | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-040-889B
|
Honeywell™
BP1201M |
Medium | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
19-040-889C
|
Honeywell™
BP1201L |
Large | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Ideal for welding and other high-heat operations
- Meets NIOSH 42 CFR Part 84 standards
Convenient backpack design allows cartridges and filters to be located on the user's back and out of the work zone—ideal for environments with high heat or sparks, such as welding, coke and smelting operations. Simply attach the backpack to North 7700 half mask cartridge connectors, then fasten the appropriate cartridges or filters to the backpack. No modifications, tools, or extra parts are needed.
Complete assembly includes backpack adapter and half-mask. Backpack adapter can also be purchased separately (below) to convert existing North facepieces to BP1000 Series air-purifying respirators
Specifications
NIOSH 42 CFR Part 84 | |
Welding and other high-heat operations | |
North™ 5500, 7700, 5400 and 7600 Series facepieces | |
Backpack adapter |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.