Learn More
Honeywell™ North™ Lightning™ Series Safety Eyewear Replacement Lenses
Frameless Lightning Plus safety glasses offer full-view unilens construction. Interchangable lenses attach to temples that provide a secure fit. Prescription insert simplifies your safety program, allowing use by your entire workforce.
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
19-130-0769
|
Honeywell™
472RL |
Mirror | N/A | N/A | N/A | ||||
19-130-0766
|
Honeywell™
469RL |
Clear | N/A | N/A | N/A | ||||
19-130-0767
|
Honeywell™
470RL |
Smoke | N/A | N/A | N/A | ||||
Description
- Anti-scratch, anti-UV 99.9% and anti-static lenses
- Available in clear, smoke or mirrored tints
- Meets ANSI Z87.1-2010 and tested to meet the requirements of the CSA Z94.3 standard
Specifications
Scratch-resistant | |
Replacement lens |