Learn More
Honeywell™ North™ Skin Cream and Conditioner
Catalog No. 19027429
Click to view available options
Product Type:
Barrier Cream
Protective Cream
Skin Conditioner
Specially formulated from nonirritating USP and cosmetic grade ingredients to help relieve occupational dermatitis and protect, clean, and condition skin. Cream and conditioner are greaseless, not tacky, so skin remains flexible and smooth.
  • 201™ Protective Cream. Water soluble cream provides a greaseless, invisible barrier against industrial irritants. Silicone-free, cream is ideal for work with adhesives and laminates. Comes in 4 oz. (118mL) tottle. Note: Not for use with harsh acids or alkalis, or with toxic substances.
  • 212™ Skin Conditioner. Rich, lubricating, and moisturizing emulsion quickly relieves dry, itchy skin. Greaseless, ideal for working with adhesives and laminates. Comes in 4 oz. (118mL) tottle.
  • 222™ Barrier Cream with Silicone. Protects skin against irritants in both wet or dry work situations. Contains silicone. Comes in 4 oz. (118mL) tottle. Note:Not for use with harsh acids or alkalis or with toxic substances.
Compliance
All are USDA authorized for use in restrooms and dressing rooms.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Barrier Cream
Certifications/Compliance USDA
For Use With (Application) Agriculture, chemical, construction, fire protection, manufacturing, medical, municipal services, nuclear, oil and gas, pharmaceutical, Steel and Metals
Packaging Bottle
Quantity 1 Ea.
Volume (English) 4 oz.
Volume (Metric) 118 mL
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.