Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOS1AP Antibody, Novus Biologicals™
SDP

Catalog No. NB436301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB436301 25 μL
NBP238151 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB436301 Supplier Novus Biologicals Supplier No. NBP23815125UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

NOS1AP Polyclonal specifically detects NOS1AP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen NOS1AP
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. O75052
Gene Alias 6330408P19Rik, CAPONMGC138500, carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein, C-terminal PDZ ligand of neuronal nitric oxide synthase protein, KIAA0464°C-terminal PDZ domain ligand of neuronal nitric oxide synthase (CAPON), ligand of neuronal nitric oxide synthase with carboxyl-terminal PDZ domain, nitric oxide synthase 1 (neuronal) adaptor protein, Nitric oxide synthase 1 adaptor protein
Gene Symbols NOS1AP
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: HTGSKVSHPQEPMLTASPRMLLPSSSSKPPGLGTETPLSTHHQMQLLQQLLQQQQQQTQVAVAQVHLLKDQLAAEAAARLEAQARVHQLLLQNKDMLQHISLLVK
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9722
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.