Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOS1AP Rabbit anti-Human, FITC, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB042423 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB042423 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB042423 Supplier Novus Biologicals Supplier No. NBP297229F

Rabbit Polyclonal Antibody

NOS1AP Polyclonal specifically detects NOS1AP in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen NOS1AP
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias 6330408P19Rik, CAPONMGC138500, carboxyl-terminal PDZ ligand of neuronal nitric oxide synthase protein, C-terminal PDZ ligand of neuronal nitric oxide synthase protein, KIAA0464C-terminal PDZ domain ligand of neuronal nitric oxide synthase (CAPON), ligand of neuronal nitric oxide synthase with carboxyl-terminal PDZ domain, nitric oxide synthase 1 (neuronal) adaptor protein, Nitric oxide synthase 1 adaptor protein
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NOS1AP fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9722
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.