Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOSTRIN Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB164619 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164619 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB164619 Supplier Novus Biologicals Supplier No. NBP281749AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NOSTRIN Polyclonal antibody specifically detects NOSTRIN in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen NOSTRIN
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias BM247 homolog, DaIP2, endothelial nitric oxide synthase traffic inducer, ENOS traffic inducer, eNOS-trafficking inducer, Nitric oxide synthase traffic inducer, nitric oxide synthase traffickerMGC20702, nitric oxide synthase trafficking, nostrin, ortholog of mouse disabled 2 interacting proein 2
Host Species Rabbit
Immunogen NOSTRIN antibody was raised against a 17 amino acid peptide near the carboxy terminus of human NOSTRIN. The immunogen is located within amino acids 430 - 480 of NOSTRIN. Amino Acid Squence: SFQARQDDELNLEKGDI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 115677
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.