Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Notch-1 Antibody (mN1A) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 7 publications
Supplier: Novus Biologicals NB10078486SS
Description
Notch-1 Monoclonal specifically detects Notch-1 in Human, Mouse, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready, Knockout Validated.Specifications
Notch-1 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1 - 2 ug/mL, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunoprecipitation 1:10 - 1:100, Immunohistochemistry-Paraffin 1:10 - 1:500, CyTOF-ready, Knockout Validated | |
EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
Mouse | |
272.505 kDa | |
0.025 mL | |
Cancer Stem Cells, Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
4851 | |
Human, Mouse, Rat (Negative) | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
mN1A | |
Unconjugated | |
Q01705 | |
NOTCH1 | |
Mouse Notch1 protein [Uniprot: Q01705] | |
Protein G purified | |
RUO | |
Primary | |
Does not cross-react with Notch 2, 3, or 4. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction