Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Notch-1 Antibody (mN1A) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 7 publications

$243.50 - $562.50

Spécifications

Antigen Notch-1
Clone mN1A
Concentration 1.0 mg/mL
Dilution Western Blot 1 - 2 ug/mL, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunoprecipitation 1:10 - 1:100, Immunohistochemistry-Paraffin 1:10 - 1:500, CyTOF-ready, Knockout Validated
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
NB10078486T
SDP
Afficher les documents
Novus Biologicals
NB10078486SS
0.025 mL
Each for $243.50
Il en reste null
ajouter au panier
 
NB10078486
SDP
Afficher les documents
Novus Biologicals
NB10078486
0.1 mL
Each for $562.50
Il en reste null
ajouter au panier
 
Description

Description

Notch-1 Monoclonal specifically detects Notch-1 in Human, Mouse, Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, CyTOF-ready, Knockout Validated.
Spécifications

Spécifications

Notch-1
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Cancer Stem Cells, Golgi Apparatus Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction
Q01705
4851
Mouse Notch1 protein [Uniprot: Q01705]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
272.505 kDa
mN1A
Western Blot 1 - 2 ug/mL, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunoprecipitation 1:10 - 1:100, Immunohistochemistry-Paraffin 1:10 - 1:500, CyTOF-ready, Knockout Validated
Monoclonal
Purified
RUO
Human, Mouse, Rat (Negative)
EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1
NOTCH1
IgG1 κ
Protein G purified
Does not cross-react with Notch 2, 3, or 4.
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications

For Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'